Learn More
Medchemexpress LLC Betaine | 107-43-7 | 99.8% | 100 G
SDP
Catalog No. 5000407069
Encompass_Preferred
Betaine (Trimethylglycine) is a natural compound and active methyl-donor that helps maintain normal DNA methylation patterns. Found ubiquitously in plants, animals, and microorganisms, it acts as an osmolyte, regulating cellular water and ion balance. This compound supports various intestinal microbes and has demonstrated inhibition of HeLa cell proliferation in vitro, as well as attenuation of liver-to-body weight ratio in vivo.
  • Maintains normal DNA methylation patterns.
  • Regulates cellular water and ion balance.
  • Improves capacity against heat stress.
  • Promotes intestinal microbial fermentation activity.
  • Inhibits proliferation of HeLa cells (in vitro).
  • Attenuates liver-to-body weight ratio (in vivo).
Catalog No. 50-004-07069 Supplier Medchemexpress LLC Supplier No. HYB0710100G
Betaine (Trimethylglycine) is a natural compound and active methyl-donor that helps maintain normal DNA methylation patterns. Found ubiquitously in plants, animals, and microorganisms, it acts as an osmolyte, regulating cellular water and ion balance. This compound supports various intestinal microbes and has demonstrated inhibition of HeLa cell proliferation in vitro, as well as attenuation of liver-to-body weight ratio in vivo.
  • Maintains normal DNA methylation patterns.
  • Regulates cellular water and ion balance.
  • Improves capacity against heat stress.
  • Promotes intestinal microbial fermentation activity.
  • Inhibits proliferation of HeLa cells (in vitro).
  • Attenuates liver-to-body weight ratio (in vivo).