Learn More
As One International Inc Betaine Enhancer solution 5 M, 5 x 1 ml
Supplier:  As One International Inc AO351104
Encompass
Betaine Enhancer Solution is an excellent additive over a wide range of different templates, including GC-rich sequences and templates with a high degree of secondary structures. Betaine binds preferentially to AT rich sequences in the major groove, thereby stabilizing AT rich regions of the DNA. Because AT forms 2 hydrogen bonds and GC forms 3, the bonding of AT is less stable than the one of GC. As a consequence of the stabilizing effect of Betaine on AT bonding, the stability of AT bonding and GC bonding is brought close to an equal level. At the same time, Betaine has a sequence independent destabilizing effect on all DNA. The Tm of AT rich and GC rich sequences are equalized and the overall Tm is lowered. Furthermore, Betaine aids the processivity of thermostable polymerases and reduces "pauses" in polymerization caused by secondary structure that can induce the polymerase to disassociate from the DNA strand.
Catalog No. 50-165-6009