Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Betaine, 98%, for analysis, anhydrous, ACROS Organics™

Click to view available options
Quantity:
5g
100g
500g
2.5kg
Packaging:
Glass Bottle
Plastic bottle
107-43-7
C5H11NO2
117.15
MFCD00012123
KWIUHFFTVRNATP-UHFFFAOYSA-N
betaine,glycine betaine,oxyneurine,lycine,trimethylglycine,abromine,trimethylglycocoll,glycylbetaine,glycocoll betaine,acidin-pepsin
247
CHEBI:17750
2-(trimethylazaniumyl)acetate
C[N+](C)(C)CC(=O)[O-]
107-43-7
C5H11NO2
117.15
MFCD00012123
KWIUHFFTVRNATP-UHFFFAOYSA-N
betaine,glycine betaine,oxyneurine,lycine,trimethylglycine,abromine,trimethylglycocoll,glycylbetaine,glycocoll betaine,acidin-pepsin
247
CHEBI:17750
2-(trimethylazaniumyl)acetate
C[N+](C)(C)CC(=O)[O-]
For analysis
Sulfate 200ppm max
Ammonium 200ppm max
Chloride 200ppm max
Transmittance 95% min. (c=10, H2O, 430nm) 1cm cell
Packaging Glass Bottle
Sulfated Ash 0.1% max.
Color White
Melting Point 301.0°C
Infrared Spectrum Authentic
Quantity 5g
Loss on Drying 1% max. (140°C, 3 hrs)
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula (CH3)3NCH2CO2
Beilstein 04, IV, 2369
Merck Index 15, 1181
Solubility Information Solubility in water: 1000g/L (25°C). Other solubilities: 55g/100 g methanol,8.7g/100 g ethanol,sparingly soluble in ether,slightly soluble in chloroform
Formula Weight 117.15
Physical Form Crystals or Crystalline Powder
Percent Purity 98%
Grade Analysis
Chemical Name or Material βine, Anhydrous
Show More Show Less

EINECSNumber : 203-490-6