Learn More
Bioworld Betaine, Anhydrous, 100 g
Supplier:  Bioworld 402000361
Encompass_Preferred
Betaine can be used to treat high homocysteine levels. Its alkylated derivates are used as quaternary ammonium zwitterionic surfactants. Also known as Glycine Betaine or Oxyneurine.Appearance: white Crystalline PowderBoiling Point : 391.94 °C (Predicted)Density : 1.00 g/mL at 20 ºCIUPAC_Name: 2-(Trimethylazaniumyl)AcetateMelting Point: 301-305 °CRTECS Number: DS5900000kg
Catalog No. 50-255-390