Learn More
Betaine, anhydrous, 98%
Description
Betaine is an active ingredient in toothpaste to control the symptoms of dryness of the mouth. It is used to treat homocystinuria, which is a defect in the major pathway of methionine biosynthesis. It is also used for boosting the immune system and for improving athletic performance. It is helpful to prevent noncancerous tumors in the colon (colorectal adenomas).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Betaine is an active ingredient in toothpaste to control the symptoms of dryness of the mouth. It is used to treat homocystinuria, which is a defect in the major pathway of methionine biosynthesis. It is also used for boosting the immune system and for improving athletic performance. It is helpful to prevent noncancerous tumors in the colon (colorectal adenomas).
Solubility
Soluble in water and methanol.
Notes
Hygroscopic. Store in cool place. Incompatible with strong oxidizing agents.
Specifications
Specifications
| Melting Point | >300°C (decomposition) |
| Assay Percent Range | 0.98 |
| Quantity | 2500 g |
| Beilstein | 3537113 |
| Sensitivity | Hygroscopic |
| Merck Index | 14,1179 |
| Solubility Information | Soluble in water and methanol. |
| Formula Weight | 117.15 |
| Percent Purity | 98% |
| Chemical Name or Material | Betaine, anhydrous |
Safety and Handling
| EINECSNumber | 203-490-6 |
| RTECSNumber | DS5900000 |
| TSCA | Yes |
| storageNote1 | Hygroscopic |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.