Learn More
Medchemexpress LLC Betaine-d3 chloride | 1219349-47-9 | 156.63 | 1 MG
SDP
Catalog No. 5000416291
Encompass_Preferred
Betaine-d3 (chloride) is a deuterium-labeled version of Betaine chloride. It is intended for research use only and not for sale to patients. Stable heavy isotopes, such as deuterium, are incorporated into drug molecules primarily as tracers for quantitative analysis during drug development, and deuterium substitution can influence the pharmacokinetic and metabolic profiles of drugs.
  • Deuterium labeled Betaine chloride.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Purity: 98.63%.
Catalog No. 50-004-16291 Supplier Medchemexpress LLC Supplier No. HYN0739S1MG
Betaine-d3 (chloride) is a deuterium-labeled version of Betaine chloride. It is intended for research use only and not for sale to patients. Stable heavy isotopes, such as deuterium, are incorporated into drug molecules primarily as tracers for quantitative analysis during drug development, and deuterium substitution can influence the pharmacokinetic and metabolic profiles of drugs.
  • Deuterium labeled Betaine chloride.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Purity: 98.63%.