Learn More
Medchemexpress LLC Betaine-d3 chloride | 1219349-47-9 | 156.63 | 5 MG
SDP
Catalog No. 5000416367
Encompass_Preferred
Betaine-d3 chloride is the deuterium labeled Betaine chloride. It is for research use only and not sold to patients.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-16367 Supplier Medchemexpress LLC Supplier No. HYN0739S5MG
Add to Cart
Add to Cart
Betaine-d3 chloride is the deuterium labeled Betaine chloride. It is for research use only and not sold to patients.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.