Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Betaine-d3 chloride | 1219349-47-9 | 98.6% | 156.63 g/mol | C5H9D3ClNO2 | 10 MG
SDP

Catalog No. 5000296042
Encompass_Preferred

Betaine-d3 chloride is the deuterium-labeled form of betaine chloride intended for use as an isotopic tracer and internal standard in quantitative NMR, GC-MS, and LC-MS analyses for research applications.

  • Deuterium-labeled internal standard for quantitative NMR, GC-MS, and LC-MS.
  • Reported purity 98.6%.
  • Molecular weight 156.63 g/mol.
  • Molecular formula C5H9D3ClNO2.
  • Available in a 10 mg package suitable for analytical workflows.
  • For research use only; not for human or clinical use.

Catalog No. 50-002-96042 Supplier Medchemexpress LLC Supplier No. HYN0739S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Betaine-d3 chloride is the deuterium-labeled form of betaine chloride intended for use as an isotopic tracer and internal standard in quantitative NMR, GC-MS, and LC-MS analyses for research applications.

  • Deuterium-labeled internal standard for quantitative NMR, GC-MS, and LC-MS.
  • Reported purity 98.6%.
  • Molecular weight 156.63 g/mol.
  • Molecular formula C5H9D3ClNO2.
  • Available in a 10 mg package suitable for analytical workflows.
  • For research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.