Learn More
Medchemexpress LLC Betaine-d9 chloride | 285979-85-3 | 162.66 | 100 MG
SDP
Catalog No. 5000416208
Encompass_Preferred
Betaine-d9 chloride is the deuterium labeled Betaine chloride. This compound is for research use only.
  • Can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-16208 Supplier Medchemexpress LLC Supplier No. HYN0739S1100MG
Betaine-d9 chloride is the deuterium labeled Betaine chloride. This compound is for research use only.
  • Can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs.