Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Betaine-d9 chloride | 285979-85-3 | 162.66 | 25 MG
SDP

Catalog No. 5000389889
Encompass_Preferred

Betaine-d9 chloride is a deuterium labeled Betaine chloride intended for research use only.

  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements are incorporated for quantitation during drug development.
  • Deuteration has the potential to affect pharmacokinetic and metabolic profiles of drugs.
  • Purity of 99.66%.
  • Appears as a white to off-white solid.
  • Recommended storage at 4°C, sealed, away from moisture.
  • For solutions, store at -80°C for 6 months or -20°C for 1 month, sealed and away from moisture.

Catalog No. 50-003-89889 Supplier Medchemexpress LLC Supplier No. 25MGHYN0739S1
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Betaine-d9 chloride is a deuterium labeled Betaine chloride intended for research use only.

  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements are incorporated for quantitation during drug development.
  • Deuteration has the potential to affect pharmacokinetic and metabolic profiles of drugs.
  • Purity of 99.66%.
  • Appears as a white to off-white solid.
  • Recommended storage at 4°C, sealed, away from moisture.
  • For solutions, store at -80°C for 6 months or -20°C for 1 month, sealed and away from moisture.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.