Learn More
Medchemexpress LLC Betaine-d9 chloride | 285979-85-3 | 162.66 | 5 MG
SDP
Catalog No. 5000417105
Encompass_Preferred
Betaine-d9 (chloride) is the deuterium labeled Betaine chloride. This compound can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-17105 Supplier Medchemexpress LLC Supplier No. HYN0739S15MG
Betaine-d9 (chloride) is the deuterium labeled Betaine chloride. This compound can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.