Learn More
Bioworld Betaine Hydrochloride, 1 kg
Catalog No. 50255389
Encompass_Preferred
Itis a non-essential nutrient and a source of hydrochloric acid, a naturally occurring chemical in the stomach that helps digest food by breaking up fats and proteins. In particular, Betaine Hydrochloride is necessary for adequate absorption of protein, calcium, vitamin B12 and iron. It is also known as hydrochloric acid and stomach acid.Ultra Pure Grade Heavy Metals(%)- <=0.0010 Identification (IR)- Pass Loss on Drying(%)- <=0.5 Residue on Ignition(%)- <=0.1Appearance: Colorless to White CrystalsDensity : 1.00 g/mlHS Code : 29239000IUPAC_Name: Carboxymethyl(Trimethyl)azanium ChlorideMelting Point: 301-305 °CPH Value: pH (25%, Water) @25C- 0.8-1.2RTECS Number: BP3136000
Catalog No. 50-255-389 Supplier Bioworld Supplier No. 402000321
Itis a non-essential nutrient and a source of hydrochloric acid, a naturally occurring chemical in the stomach that helps digest food by breaking up fats and proteins. In particular, Betaine Hydrochloride is necessary for adequate absorption of protein, calcium, vitamin B12 and iron. It is also known as hydrochloric acid and stomach acid.Ultra Pure Grade Heavy Metals(%)- <=0.0010 Identification (IR)- Pass Loss on Drying(%)- <=0.5 Residue on Ignition(%)- <=0.1Appearance: Colorless to White CrystalsDensity : 1.00 g/mlHS Code : 29239000IUPAC_Name: Carboxymethyl(Trimethyl)azanium ChlorideMelting Point: 301-305 °CPH Value: pH (25%, Water) @25C- 0.8-1.2RTECS Number: BP3136000