Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Betaine hydrochloride, 99%

Catalog No. AAA161220B
Change view
Click to view available options
Quantity:
250 g
1000 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA161220B 1000 g
AAA1612230 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA161220B Supplier Thermo Scientific Chemicals Supplier No. A161220B
Only null left
Add to Cart

CAS: 590-46-5 | C5H12ClNO2 | 153.61 g/mol

Betaine hydrochloride is a naturally occurring compound that has been of interest for its role in osmoregulation. As a drug, it has been used as a source of hydrochloric acid in the treatment of hypochlorhydria. Betaine has also been used in the treatment of liver disorders, for hyperkalemia, for homocystinuria and for gastrointestinal disturbances.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Betaine hydrochloride is a naturally occurring compound that has been of interest for its role in osmoregulation. As a drug, it has been used as a source of hydrochloric acid in the treatment of hypochlorhydria. Betaine has also been used in the treatment of liver disorders, for hyperkalemia, for homocystinuria and for gastrointestinal disturbances.

Solubility
Soluble in water.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 590-46-5
Molecular Formula C5H12ClNO2
Molecular Weight (g/mol) 153.61
MDL Number MFCD00011903
InChI Key HOPSCVCBEOCPJZ-UHFFFAOYSA-N
Synonym betaine hydrochloride, betaine chloride, pluchine, acidogeno, acipepsol, achylin, acidin, acinorm, acidol, acidine
PubChem CID 11545
SMILES [Cl-].C[N+](C)(C)CC(O)=O

Specifications

Melting Point ∼240°C (decomposition)
Quantity 1000 g
Beilstein 3916181
Merck Index 14,1179
Solubility Information Soluble in water.
IUPAC Name (carboxymethyl)trimethylazanium chloride
Formula Weight 153.61
Percent Purity 99%
Chemical Name or Material Betaine hydrochloride
Hazard Category H319
Hazard Statement GHS H Statement
H319
Causes serious eye irritation.
Precautionary Statement P264b-P280i-P305+P351+P338
EINECSNumber 209-683-1
RTECSNumber BP3136000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.