Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Betaine (Trimethylglycine) | 107-43-7 | 100.0% | 25 G
SDP

Catalog No. 5000405586
Encompass_Preferred

Betaine (Trimethylglycine) is a natural compound and active methyl-donor, maintaining normal DNA methylation patterns. Ubiquitous in plants, animals, and microorganisms, it functions as an osmolyte, balancing cellular water and ions. This promotes intestinal microbes against osmotic variations, enhancing fermentation activity.

  • Purity of 99.97%
  • Appears as a solid
  • Color is white to off-white
  • Initial source includes endogenous metabolites, disease markers, cancer, plants, animals, and microorganisms
  • Structure classified as others

Catalog No. 50-004-05586 Supplier Medchemexpress LLC Supplier No. 25GHYB0710
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Betaine (Trimethylglycine) is a natural compound and active methyl-donor, maintaining normal DNA methylation patterns. Ubiquitous in plants, animals, and microorganisms, it functions as an osmolyte, balancing cellular water and ions. This promotes intestinal microbes against osmotic variations, enhancing fermentation activity.

  • Purity of 99.97%
  • Appears as a solid
  • Color is white to off-white
  • Initial source includes endogenous metabolites, disease markers, cancer, plants, animals, and microorganisms
  • Structure classified as others

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.