Learn More
Biomatik Corporation Beta-Lactoglobulin (Blg) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU29955
Encompass_Preferred
Beta-Lactoglobulin (Blg) Polyclonal Antibody is a Bovine (Bos taurus) Polyclonal Antibody to Beta-Lactoglobulin (bLg). The Beta-Lactoglobulin (Blg) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Bovine (Bos taurus). Applications: WB, IHC, ICC, IP. Target Synonyms: Beta-Lactoglobulin (bLg). Accession Number: P02754. Gene ID: 280838. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7263