Learn More
Cayman Chemical Betamethasone 17-valerate, Purity: ≥98%, CAS: 2152-44-5, 100 mg
SDP
Catalog No. 502936742
Encompass_Preferred
A glucocorticoid with anti-inflammatory activity; inhibits the binding of the radiolabeled glucocorticoid dexamethasone to human epidermis and mouse skin (IC50s = 5 and 6 nM, respectively); topical application of a 0.12% ointment blocks increases in ear weight in a rat model of ear edema induced by croton oil
Catalog No. 50-293-6742 Supplier Cayman Chemical Supplier No. 25640100 MG
A glucocorticoid with anti-inflammatory activity; inhibits the binding of the radiolabeled glucocorticoid dexamethasone to human epidermis and mouse skin (IC50s = 5 and 6 nM, respectively); topical application of a 0.12% ointment blocks increases in ear weight in a rat model of ear edema induced by croton oil