Learn More
Cayman Chemical Betamethasone, Purity: ≥98%, CAS: 378-44-9, 50 mg
SDP
Catalog No. 502983593
Encompass_Preferred
A synthetic corticosteroid; reduces BALF MMP-2 and MMP-9 activities, TNF-α levels, and neutrophil number in a mouse model of LPS-induced lung injury; prenatal administration improves neonatal lung function but decreases hippocampal and corpus callosum neuron density and induces motor function deficits in a rabbit model of late preterm birth at 0.1 mg/kg; extends gestational length in a rat model of HMGB1-induced preterm labor
Catalog No. 50-298-3593 Supplier Cayman Chemical Supplier No. 2036350 MG
A synthetic corticosteroid; reduces BALF MMP-2 and MMP-9 activities, TNF-α levels, and neutrophil number in a mouse model of LPS-induced lung injury; prenatal administration improves neonatal lung function but decreases hippocampal and corpus callosum neuron density and induces motor function deficits in a rabbit model of late preterm birth at 0.1 mg/kg; extends gestational length in a rat model of HMGB1-induced preterm labor