Learn More
Cayman Chemical Betamethasone Dipropionate, Purity: ≥98%, CAS: 5593-20-4, 100 mg
SDP
Catalog No. 502917586
Encompass_Preferred
A synthetic corticosteroid and prodrug from of betamethasone; binds to the glucocorticoid receptor (IC50 = 5.25 nM); decreases the number of circulating eosinophils in dogs; reduces croton oil-induced exudate formation in a granuloma pouch assay in rats
Catalog No. 50-291-7586 Supplier Cayman Chemical Supplier No. 39499100 MG
A synthetic corticosteroid and prodrug from of betamethasone; binds to the glucocorticoid receptor (IC50 = 5.25 nM); decreases the number of circulating eosinophils in dogs; reduces croton oil-induced exudate formation in a granuloma pouch assay in rats