Learn More
Medchemexpress LLC HY-13570 100mg Medchemexpress, Betamethasone CAS:378-44-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY13570100MG
Encompass_Preferred
Medchemexpress, HY-13570 100mg Betamethasone CAS:378-44-9 Betamethasone is a synthetic glucocorticoid with anti-inflammatory and immunosuppressive activities. Betamethasone accelerates fetal lung maturation and induces gene expression and apoptosis[4]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1398