Learn More
Apexbio Technology LLC Betamethasone 378-44-9 100mg
SDP
Catalog No. 502240334
Encompass_Preferred
Betamethasone is a synthetic corticosteroid functioning primarily as a glucocorticoid receptor agonist Mechanistically it modulates gene expression through binding glucocorticoid receptors regulating transcription factors and subsequently affecting protein synthesis Betamethasone influences inflammatory pathways by inhibiting key enzyme expressions such as phospholipase A2 cyclooxygenase-2 (COX-2) and nitric oxide synthase (NOS) resulting in decreased synthesis of inflammatory mediators like prostaglandins leukotrienes and nitric oxide Additionally it has been reported to suppress keratinocyte proliferation Due to its glucocorticoid-modulating properties and anti-inflammatory effects betamethasone is commonly employed in biomedical research involving inflammatory signaling pathways dermatological models and gene expression studies
This item is not returnable. View return policy
Catalog No. 50-224-0334 Supplier Apexbio Technology LLC Supplier No. B1896100
Betamethasone is a synthetic corticosteroid functioning primarily as a glucocorticoid receptor agonist Mechanistically it modulates gene expression through binding glucocorticoid receptors regulating transcription factors and subsequently affecting protein synthesis Betamethasone influences inflammatory pathways by inhibiting key enzyme expressions such as phospholipase A2 cyclooxygenase-2 (COX-2) and nitric oxide synthase (NOS) resulting in decreased synthesis of inflammatory mediators like prostaglandins leukotrienes and nitric oxide Additionally it has been reported to suppress keratinocyte proliferation Due to its glucocorticoid-modulating properties and anti-inflammatory effects betamethasone is commonly employed in biomedical research involving inflammatory signaling pathways dermatological models and gene expression studies