Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Betamethasone 17-propionate | 5534-13-4 | MFCD19705255 | >98.0% | 448.52 | C25H33FO6 | 10 MG
SDP

Catalog No. 5000282406
Encompass_Preferred

Betamethasone 17-propionate is the 17-propionate ester of the glucocorticoid betamethasone, supplied for research use in preclinical studies and analytical applications. It is commonly used to investigate anti-inflammatory effects, pharmacology, and as a reference standard in bioanalytical and toxicology workflows.

  • High purity suitable for research (>98.0%).
  • Used in anti-inflammatory and pharmacology studies.
  • Available in small research sizes (5 mg, 10 mg; larger quantities by inquiry).
  • Molecular formula C25H33FO6.
  • Molecular weight 448.52 g·mol-1.
  • CAS number 5534-13-4 for unambiguous identification.

Catalog No. 50-002-82406 Supplier Medchemexpress LLC Supplier No. HYW34245810MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Betamethasone 17-propionate is the 17-propionate ester of the glucocorticoid betamethasone, supplied for research use in preclinical studies and analytical applications. It is commonly used to investigate anti-inflammatory effects, pharmacology, and as a reference standard in bioanalytical and toxicology workflows.

  • High purity suitable for research (>98.0%).
  • Used in anti-inflammatory and pharmacology studies.
  • Available in small research sizes (5 mg, 10 mg; larger quantities by inquiry).
  • Molecular formula C25H33FO6.
  • Molecular weight 448.52 g·mol-1.
  • CAS number 5534-13-4 for unambiguous identification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.