Learn More
Medchemexpress LLC Betamethasone 17-propionate | 5534-13-4 | MFCD19705255 | 98.2% | 448.52 g·mol⁻¹ | C25H33FO6 | 5 MG
SDP
Catalog No. 5000282196
Encompass_Preferred
Betamethasone 17-propionate is a synthetic corticosteroid provided as a research-grade reagent and analytical standard. It is a white to off-white solid (CAS 5534-13-4; MW 448.52 g·mol⁻¹) commonly used in pharmacology and biochemical assays to study glucocorticoid activity. Powder is stable when stored at -20°C and is supplied in milligram-scale quantities for analytical work.
  • High purity suitable for analytical and research use.
  • White to off-white solid, stable when stored at -20°C.
  • Supplied with datasheet and certificate of analysis for lot-specific data.
  • Suitable for biochemical assays and glucocorticoid-related studies.
  • Available in milligram-scale quantities for reference and testing.
Catalog No. 50-002-82196 Supplier Medchemexpress LLC Supplier No. HYW3424585MG
Betamethasone 17-propionate is a synthetic corticosteroid provided as a research-grade reagent and analytical standard. It is a white to off-white solid (CAS 5534-13-4; MW 448.52 g·mol⁻¹) commonly used in pharmacology and biochemical assays to study glucocorticoid activity. Powder is stable when stored at -20°C and is supplied in milligram-scale quantities for analytical work.
  • High purity suitable for analytical and research use.
  • White to off-white solid, stable when stored at -20°C.
  • Supplied with datasheet and certificate of analysis for lot-specific data.
  • Suitable for biochemical assays and glucocorticoid-related studies.
  • Available in milligram-scale quantities for reference and testing.