Learn More
Selleck Chemical LLC Betamethasone S1500-50mg
SDP
Catalog No. 503108333
Encompass_Preferred
Betamethasone (NSC-39470 SCH-4831) is a glucocorticoid steroid with anti-inflammatory and immunosuppressive properties
Catalog No. 50-310-8333 Supplier Selleck Chemical LLC Supplier No. S150050MG
Betamethasone (NSC-39470 SCH-4831) is a glucocorticoid steroid with anti-inflammatory and immunosuppressive properties