Learn More
Medchemexpress LLC Betamethasone acetate | 987-24-6 | MFCD00867462 | 99.4% | 434.50 g/mol | C24H31FO6 | 5 MG
SDP
Catalog No. 5000222464
Encompass_Preferred
Betamethasone acetate is a synthetic glucocorticoid corticosteroid used as a research reagent and analytical standard in pharmacological and biochemical studies. It exhibits anti-inflammatory and immunosuppressive activity and is typically provided in small, measured quantities for laboratory assays.
  • High purity appropriate for analytical and research applications.
  • Anti-inflammatory and immunosuppressive activity for pharmacology studies.
  • Available in small, measured quantities suitable for bench experiments.
  • Supplied with supporting documentation such as COA and SDS.
  • Soluble in common organic solvents for assay preparation.
Catalog No. 50-002-22464 Supplier Medchemexpress LLC Supplier No. HY13570B5MG
Betamethasone acetate is a synthetic glucocorticoid corticosteroid used as a research reagent and analytical standard in pharmacological and biochemical studies. It exhibits anti-inflammatory and immunosuppressive activity and is typically provided in small, measured quantities for laboratory assays.
  • High purity appropriate for analytical and research applications.
  • Anti-inflammatory and immunosuppressive activity for pharmacology studies.
  • Available in small, measured quantities suitable for bench experiments.
  • Supplied with supporting documentation such as COA and SDS.
  • Soluble in common organic solvents for assay preparation.