Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Betamethasone Dipropionate, Micronized Powder, USP, 97-103%, Spectrum™ Chemical
SDP

Catalog No. 18600946 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
1 kg
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-600-946 1 g Amber Glass Bottle
18-600-947 5 g Amber Glass Bottle
18-600-948 25 g Amber Glass Bottle
18-600-950 100 g Amber Glass Bottle
18-600-949 1 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-600-946 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. BE1811GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

BE181, 5593-20-4, C28H37FO7

Spectrum™ Chemical Betamethasone Dipropionate, Micronized Powder, USP is a glucocorticoid steroid applied as a topical cream, ointment, lotion or gel to treat itching and other minor skin conditions. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 5593-20-4
Molecular Formula C28H37FO7
Molecular Weight (g/mol) 504.60
InChI Key CIWBQSYVNNPZIQ-XYWKZLDCSA-N
IUPAC Name 2-[(1R,2S,3aS,3bS,9aS,9bR,10S,11aS)-9b-fluoro-10-hydroxy-2,9a,11a-trimethyl-7-oxo-1-(propanoyloxy)-1H,2H,3H,3aH,3bH,4H,5H,7H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-yl]-2-oxoethyl propanoate
SMILES CCC(=O)OCC(=O)[C@@]1(OC(=O)CC)[C@@H](C)C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]12C

Specifications

Packaging Amber Glass Bottle
CAS Max % 100%
Industry Type Pharmaceutical
Quantity 1 g
Percent Purity 97 to 103%
Grade USP
Chemical Name or Material Betamethasone Dipropionate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.