Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Betamethasone Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111014091 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4091 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4091 Supplier MilliporeSigma Supelco Supplier No. PHR13981G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.Betamethasone belongs to the corticosteriod family of active pharmaceutical ingredients (APIs). It exhibits anti-inflammatory activity and hence is used in the manufacture of various finished pharmaceutical products and is also employed as a starting material to manufacture other APIs that are related to this steroid family. Featured Industries: Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

CAS 378-44-9
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Physical Form Neat
Linear Formula C22H29 FO5
MDL Number MFCD00062969
Melting Point 235°C to 237°C
Synonym Betamethasone; 9 alpha-Fluoro-11 beta,17 alpha,21-trihydroxy-16 beta-methylpregna-1,4-diene-3,20-dione; 9 alpha-Fluoro-16 beta-methyl-11 beta,17 alpha,21-trihydroxy-1,4-pregnadiene-3,20-dione; 9 alpha-Fluoro-16 beta-methylprednisolone
Type Certified Reference Material
Quantity 1 g
Formula Weight 392.46
Grade Certified Reference Material
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.