Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Betamethasone Sodium Phosphate, USP, 97-103%, Spectrum™ Chemical
SDP

Catalog No. 18600951 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
1 kg
Packaging:
Amber Glass Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-600-951 1 g Amber Glass Bottle
18-600-952 5 g Amber Glass Bottle
18-600-953 25 g Amber Glass Bottle
18-600-954 100 g Amber Glass Bottle
18-600-955 1 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-600-951 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. BE1831GM
Add to Cart
Add to Cart

BE183, 151-73-5, C22H28FNa2O8P

Spectrum™ Chemical Betamethasone Sodium Phosphate, USP is a strong glucocorticoid steroid used to reduce inflammation, irritation, and pain. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 151-73-5
Molecular Formula C22H28FNa2O8P
Molecular Weight (g/mol) 516.41
InChI Key PLCQGRYPOISRTQ-LWCNAHDDSA-L
IUPAC Name disodium (1R,2S,3aS,3bS,9aS,9bR,10S,11aS)-9b-fluoro-1,10-dihydroxy-2,9a,11a-trimethyl-1-[2-(phosphonatooxy)acetyl]-1H,2H,3H,3aH,3bH,4H,5H,7H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-one
SMILES [Na+].[Na+].C[C@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]2(C)[C@@]1(O)C(=O)COP([O-])([O-])=O

Specifications

CAS Max % 100%
Packaging Amber Glass Bottle
Quantity 1 g
Specific Rotation +99° to +105°
Percent Purity 97 to 103%
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.