Learn More
Apexbio Technology LLC Betamethasone Valerate 2152-44-5 10mM (in 1mL DMSO)
SDP
Catalog No. 5000096123
Encompass_Preferred
Betamethasone valerate (CAS 2152-44-5) is a small-molecule agonist targeting the glucocorticoid receptor It is designed to modulate inflammatory responses by altering gene transcription thereby regulating immune-mediated processes Betamethasone valerate exerts its biological activity primarily through glucocorticoid receptor agonism In in vitro studies betamethasone valerate inhibits radiolabeled glucocorticoid (dexamethasone) binding to human epidermis and mouse skin with IC50 values of 5 and 6 nM respectively Based on these pharmacological properties betamethasone valerate holds research potential in dermatological applications including investigations of recurrent aphthous stomatitis and inflammatory skin disorders
Catalog No. 50-000-96123 Supplier Apexbio Technology LLC Supplier No. A83795.1
Betamethasone valerate (CAS 2152-44-5) is a small-molecule agonist targeting the glucocorticoid receptor It is designed to modulate inflammatory responses by altering gene transcription thereby regulating immune-mediated processes Betamethasone valerate exerts its biological activity primarily through glucocorticoid receptor agonism In in vitro studies betamethasone valerate inhibits radiolabeled glucocorticoid (dexamethasone) binding to human epidermis and mouse skin with IC50 values of 5 and 6 nM respectively Based on these pharmacological properties betamethasone valerate holds research potential in dermatological applications including investigations of recurrent aphthous stomatitis and inflammatory skin disorders