Learn More
Apexbio Technology LLC Betamipron 3440-28-6 50mg
SDP
Catalog No. 502240771
Encompass_Preferred
Betamipron (CAS 3440-28-6) is a small-molecule inhibitor targeting renal tubular uptake mechanisms It is designed to inhibit renal tubular uptake thereby reducing nephrotoxicity during carbapenem antibiotic therapy Betamipron exerts its biological activity primarily through blockade of the renal tubular uptake process Its potency is typically assessed via renal uptake assays and cellular transporter inhibition tests however exact IC50 values depend on experimental conditions and models and are not explicitly provided Based on these pharmacological properties Betamipron holds research potential in the investigation of antibiotic-related renal pharmacokinetics carbapenem distribution profiles and nephrotoxic side-effects
This item is not returnable. View return policy
Catalog No. 50-224-0771 Supplier Apexbio Technology LLC Supplier No. B167950
Betamipron (CAS 3440-28-6) is a small-molecule inhibitor targeting renal tubular uptake mechanisms It is designed to inhibit renal tubular uptake thereby reducing nephrotoxicity during carbapenem antibiotic therapy Betamipron exerts its biological activity primarily through blockade of the renal tubular uptake process Its potency is typically assessed via renal uptake assays and cellular transporter inhibition tests however exact IC50 values depend on experimental conditions and models and are not explicitly provided Based on these pharmacological properties Betamipron holds research potential in the investigation of antibiotic-related renal pharmacokinetics carbapenem distribution profiles and nephrotoxic side-effects