Learn More
Texwipe™ BetaMop™ II
SDP
Catalog No. 19099196 Shop All Texwipe Products
Change view
Click to view available options
Product Type:
BetaMop II Mop
Mop Head Refill
For Use With (Equipment):
BetaMop II
Wring-bucket Systems
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type For Use With (Equipment)
19-099-196 BetaMop II Mop Wring-bucket Systems
19-099-197 Mop Head Refill BetaMop II
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-099-196 Supplier Texwipe™ Supplier No. TX7106
Add to Cart
Add to Cart
Low contamination properties make this the perfect mop for floor cleaning in critical environments
Durable, absorbent, corrosion-resistant, and low-contaminating; Autoclavable.
Durability, corrosion resistance, and low contamination properties make this the perfect mop for floor cleaning in critical environments. High-capacity, seamless, tubular-knit polyester mop head provides excellent absorbency. BetaMop II may be used with all wring-bucket systems. Both the mop head and clamp are autoclavable for use in sterile environments.
Specifications
Product Type BetaMop II Mop
Autoclavable Autoclavable
Resistant To Corrosion
Includes Handle and Head Refill
For Use With (Equipment) Wring-bucket Systems
For Use With (Application) For floor cleaning in both electronic cleanrooms and sterile environments
Material Polyester