Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Betanin (Red Beet Extract Diluted With Dextrin)

Catalog No. 9012665G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-126-65G 5 g
90-126-61G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 90-126-65G Supplier Sigma Aldrich Supplier No. 9012665G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Betanin is a glycosidic, indoline pigment that derives from red beets. Betanin has been used as a building block for chemical synthesis, as a food additive, and in analytical, material, and biological studies. This same product was formerly listed as CDS000584, offered red beet extract diluted with dextrin. Betanin is susceptible to degradation in light and elevated temperatures. When kept dry, it is stable to oxygen.

Specifications

CAS 877-07-6
Molecular Formula C24H26N2O13
Linear Formula C24H26N2O13
Quantity 5 g
Synonym Betanidin 5-beta-D-glucopyranoside
Molecular Weight (g/mol) 550.47 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.