Learn More
Med Vet International Betasept 4% Antiseptic Surgical Scrub 16oz, 12 Count
Supplier:  Med Vet International OTCBTA016
Encompass_Preferred
For surgical hand scrubbing and patient preoperative skin preparationChlorhexidine gluconate 4% provides antiseptic action with a persistent antimicrobial effect against a range of microorganisms. Does not contain tints or dyes.Surgical hand scrub: significantly reduces the number of microorganisms on the hands and forearms prior to surgery or patient careHealthcare personnel handwash: helps reduce bacteria that potentially can cause infectionPatient preoperative skin preparation: for preparation of the patient’s skin prior to surgerySkin wound and general skin cleansing
Catalog No. 50-304-5403