Learn More
Kyfora Bio Beta-sitosterol 25 MG
Catalog No. 502559802
Encompass_Preferred
LipoVectorTM is our liposomal transfection reagent designed for the delivery of RNA DNA and proteins into eukaryotic cells (in vitro and in vivo). LipoVectorTM interacts with nucleic acids and the cell membrane to provide efficient and safe delivery into cells. It can be used for transient and stable gene expression as well as co-transfections and is a powerful tool for intracellular delivery of nucleotides.
Catalog No. 50-255-9802 Supplier Kyfora Bio Supplier No. 1449525
LipoVectorTM is our liposomal transfection reagent designed for the delivery of RNA DNA and proteins into eukaryotic cells (in vitro and in vivo). LipoVectorTM interacts with nucleic acids and the cell membrane to provide efficient and safe delivery into cells. It can be used for transient and stable gene expression as well as co-transfections and is a powerful tool for intracellular delivery of nucleotides.