Learn More
Aat Bioquest Beta-trifluoromethylumbelliferone [7-Hydroxy-4-trifluoromethylcoumarin] [CAS 575-03-1]
SDP
Catalog No. 502979290
Encompass_Preferred
This coumarin compound is used as a calibration standard for 7-hydroxy-4-trifluoromethylcoumarin-based enzyme substrates.
Catalog No. 50-297-9290 Supplier Aat Bioquest Supplier No. 82
This coumarin compound is used as a calibration standard for 7-hydroxy-4-trifluoromethylcoumarin-based enzyme substrates.