Learn More
Medchemexpress LLC Betaxolol-d5 | 1189957-99-0 | >95.0% | 1 MG
SDP
Catalog No. 5000407355
Encompass_Preferred
Betaxolol-d5 is the deuterium-labeled version of Betaxolol. Betaxolol is a selective beta1 adrenergic receptor blocker that can be utilized in research pertaining to hypertension and glaucoma. This compound can serve as a tracer and as an internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS. Stable heavy isotopes, including deuterium, have been incorporated into drug molecules primarily to act as tracers for quantitation during the drug development process. Deuteration is notable for its potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer for quantitation during drug development.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration may influence the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-07355 Supplier Medchemexpress LLC Supplier No. HYB0381S1MG
Betaxolol-d5 is the deuterium-labeled version of Betaxolol. Betaxolol is a selective beta1 adrenergic receptor blocker that can be utilized in research pertaining to hypertension and glaucoma. This compound can serve as a tracer and as an internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS. Stable heavy isotopes, including deuterium, have been incorporated into drug molecules primarily to act as tracers for quantitation during the drug development process. Deuteration is notable for its potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer for quantitation during drug development.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration may influence the pharmacokinetic and metabolic profiles of drugs.