Learn More
Apexbio Technology LLC Betaxolol HCl 63659-19-8 10mM (in 1mL DMSO)
SDP
Catalog No. 5000061909
Encompass_Preferred
Betaxolol HCl (CAS 63659-19-8) is a selective inhibitor targeting the adrenergic receptor It demonstrates antagonist activity by competitively binding and inhibiting this receptor subtype which plays a significant role in cardiovascular regulation Betaxolol exhibits an IC value of approximately 6 M indicative of moderate inhibitory potency In biomedical research betaxolol serves as a pharmacological tool to investigate receptor signaling pathways and their involvement in conditions such as hypertension arrhythmias and cardiovascular disorders
Catalog No. 50-000-61909 Supplier Apexbio Technology LLC Supplier No. B13535.1
Betaxolol HCl (CAS 63659-19-8) is a selective inhibitor targeting the adrenergic receptor It demonstrates antagonist activity by competitively binding and inhibiting this receptor subtype which plays a significant role in cardiovascular regulation Betaxolol exhibits an IC value of approximately 6 M indicative of moderate inhibitory potency In biomedical research betaxolol serves as a pharmacological tool to investigate receptor signaling pathways and their involvement in conditions such as hypertension arrhythmias and cardiovascular disorders