Learn More
Medchemexpress LLC HY-B0381A 10mg Medchemexpress, Betaxolol (hydrochloride) CAS:63659-19-8 Purity:>98%
SDP
Catalog No. 502082505
Encompass_Preferred
Medchemexpress, HY-B0381A 10mg Betaxolol (hydrochloride) CAS:63659-19-8 Betaxolol Hydrochloride is a selective beta1 adrenergic receptor blocker that can be used for the research of hypertension and glaucoma. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2505 Supplier Medchemexpress LLC Supplier No. HYB0381A10MG
Medchemexpress, HY-B0381A 10mg Betaxolol (hydrochloride) CAS:63659-19-8 Betaxolol Hydrochloride is a selective beta1 adrenergic receptor blocker that can be used for the research of hypertension and glaucoma. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.