Learn More
Medchemexpress LLC Betaxolol hydrochloride | 63659-19-8 | 98.1% | 10 MM * 1 ML
SDP
Catalog No. 5000406565
Encompass_Preferred
Betaxolol hydrochloride is a selective beta1 adrenergic receptor blocker that can be used for the research of hypertension and glaucoma. It is a cardioselective beta-adrenergic receptor blocking agent.
  • Functions as a selective beta1 adrenergic receptor blocker
  • Cardioselective beta-adrenergic receptor blocking agent
  • Applicable for research in hypertension
  • Applicable for research in glaucoma
Catalog No. 50-004-06565 Supplier Medchemexpress LLC Supplier No. HYB0381A10MM/1ML
Betaxolol hydrochloride is a selective beta1 adrenergic receptor blocker that can be used for the research of hypertension and glaucoma. It is a cardioselective beta-adrenergic receptor blocking agent.
  • Functions as a selective beta1 adrenergic receptor blocker
  • Cardioselective beta-adrenergic receptor blocking agent
  • Applicable for research in hypertension
  • Applicable for research in glaucoma