Learn More
Medchemexpress LLC Betaxolol hydrochloride | 63659-19-8 | 98.1% | 5 MG
SDP
Catalog No. 5000406566
Encompass_Preferred
Betaxolol hydrochloride is a selective beta1 adrenergic receptor blocker, useful for research into hypertension and glaucoma. It acts as a cardioselective beta-adrenergic receptor blocking agent, potentially safer for patients with reactive airway disease due to reduced systemic beta 2- and beta 1-adrenergic receptor blockade compared to other agents. In vivo studies show it can attenuate anxiety-like behavior during early withdrawal from chronic cocaine administration in rats.
  • Selective beta1 adrenergic receptor blocker
  • Used for research of hypertension and glaucoma
  • Cardioselective beta-adrenergic receptor blocking agent
  • Attenuates anxiety-like behavior during cocaine withdrawal in rats
  • Potentially safer for patients with reactive airway disease
Catalog No. 50-004-06566 Supplier Medchemexpress LLC Supplier No. HYB0381A5MG
Betaxolol hydrochloride is a selective beta1 adrenergic receptor blocker, useful for research into hypertension and glaucoma. It acts as a cardioselective beta-adrenergic receptor blocking agent, potentially safer for patients with reactive airway disease due to reduced systemic beta 2- and beta 1-adrenergic receptor blockade compared to other agents. In vivo studies show it can attenuate anxiety-like behavior during early withdrawal from chronic cocaine administration in rats.
  • Selective beta1 adrenergic receptor blocker
  • Used for research of hypertension and glaucoma
  • Cardioselective beta-adrenergic receptor blocking agent
  • Attenuates anxiety-like behavior during cocaine withdrawal in rats
  • Potentially safer for patients with reactive airway disease