Learn More
Cayman Chemical BetaxololhydrochlorIde 10mg
SDP
Catalog No. 502965651
Encompass_Preferred
A β1-AR antagonist (Ki = 23 nM); selective for the β1-AR over the β2-AR (Ki = 790 nM); cytotoxic to 10.014 pRSV-T human corneal epithelial cells; reduces intraocular pressure in cynomolgus monkeys with ocular hypertension induced by laser trabeculoplasty at 150 UG/animal; inhibits veratridine-induced sodium influx in rat cortical synaptosomes (IC50 = 28.3 µM)
Catalog No. 50-296-5651 Supplier Cayman Chemical Supplier No. 1862510 MG
A β1-AR antagonist (Ki = 23 nM); selective for the β1-AR over the β2-AR (Ki = 790 nM); cytotoxic to 10.014 pRSV-T human corneal epithelial cells; reduces intraocular pressure in cynomolgus monkeys with ocular hypertension induced by laser trabeculoplasty at 150 UG/animal; inhibits veratridine-induced sodium influx in rat cortical synaptosomes (IC50 = 28.3 µM)