Learn More
Biocare Medical, LLC Betazoid DAB Chromogen Kit - 100 ml
SDP
Catalog No. 5082374
Encompass_Preferred
Betazoid DAB is the third-generation DAB product developed by Biocare. It is superior to conventional DAB and Cardassian DAB in terms of stability and staining intensity. This chromogen is compatible with horseradish peroxidase (HRP), not soluble in alcohol or xylene, and can be coverslipped just like any other DAB. When in the presence of peroxidase enzyme, Betazoid produces a brown stain. Betazoid DAB may increase antibody titers by two fold; and can be used manually or on automated stainers.
Catalog No. 50-823-74 Supplier Biocare Medical, LLC Supplier No. BDB2004L
Betazoid DAB is the third-generation DAB product developed by Biocare. It is superior to conventional DAB and Cardassian DAB in terms of stability and staining intensity. This chromogen is compatible with horseradish peroxidase (HRP), not soluble in alcohol or xylene, and can be coverslipped just like any other DAB. When in the presence of peroxidase enzyme, Betazoid produces a brown stain. Betazoid DAB may increase antibody titers by two fold; and can be used manually or on automated stainers.