Learn More
TARGETMOL CHEMICALS INC Betazole dihydrochloride 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T859925MG
Encompass_Preferred
Also available in 1 mL, 1 mg, 5 mg, 10 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Betazole dihydrochloride (Betazole hydrochloride) is an agonist of H2 receptor.
Catalog No. 50-287-7953