Learn More
Cayman Chemical Betazole (hydrochloride), Purity: ≥95%, CAS: 138-92-1, 100 mg
SDP
Catalog No. 502943495
Encompass_Preferred
A histamine H2 receptor agonist; induces acid secretion in isolated human gastric glands (EC50 = 120 µM); increases pepsin and acid levels in gastric juice in dogs at 3-10 mg/kg
Catalog No. 50-294-3495 Supplier Cayman Chemical Supplier No. 41712100 MG
A histamine H2 receptor agonist; induces acid secretion in isolated human gastric glands (EC50 = 120 µM); increases pepsin and acid levels in gastric juice in dogs at 3-10 mg/kg