Learn More
Cayman Chemical BetazolehydrochlorIde 10mg
SDP
Catalog No. 502943378
Encompass_Preferred
A histamine H2 receptor agonist; induces acid secretion in isolated human gastric glands (EC50 = 120 µM); increases pepsin and acid levels in gastric juice in dogs at 3-10 mg/kg
Catalog No. 50-294-3378 Supplier Cayman Chemical Supplier No. 4171210 MG
A histamine H2 receptor agonist; induces acid secretion in isolated human gastric glands (EC50 = 120 µM); increases pepsin and acid levels in gastric juice in dogs at 3-10 mg/kg