Learn More
Cayman Chemical Betazole (hydrochloride), Purity: ≥95%, CAS: 138-92-1, 50 mg
SDP
Catalog No. 502943144
Encompass_Preferred
A histamine H2 receptor agonist; induces acid secretion in isolated human gastric glands (EC50 = 120 µM); increases pepsin and acid levels in gastric juice in dogs at 3-10 mg/kg
Catalog No. 50-294-3144 Supplier Cayman Chemical Supplier No. 4171250 MG
A histamine H2 receptor agonist; induces acid secretion in isolated human gastric glands (EC50 = 120 µM); increases pepsin and acid levels in gastric juice in dogs at 3-10 mg/kg