Learn More
Enterprise Technology Solutions Betco Corporation Air Freshener,Sentec,Conc,FastDraw 46,2L,2/CT,Amber
SDP
Catalog No. 5000832504
Encompass_Preferred
Sentec Odor Eliminator traps and permanently removes odors from the air, significantly reducing the malodor present. Unique, dual malodor-counteractant technology eliminates odors and leaves behind the fresh, high-end fragrance of Mountain Meadow. Concentrated formula is effective against a broad range of odors. It meets and exceeds California VOC content requirements. Dilution ratios range from 1:10 to 1:256 according to application. As part of the FastDraw system, it is numbered (FastDraw 46) and color-coded to eliminate misuse. Spill-proof bottle helps prevent chemical contact with cleaning staff. Patented technology uses two metering tips for low-flow and high-flow dilution. Curved siphon tubes on the FastDraw Dispensers (not included) allow complete product evacuation, eliminating waste.
Catalog No. 50-008-32504 Supplier Enterprise Technology Solutions Supplier No. BET4125B200
Sentec Odor Eliminator traps and permanently removes odors from the air, significantly reducing the malodor present. Unique, dual malodor-counteractant technology eliminates odors and leaves behind the fresh, high-end fragrance of Mountain Meadow. Concentrated formula is effective against a broad range of odors. It meets and exceeds California VOC content requirements. Dilution ratios range from 1:10 to 1:256 according to application. As part of the FastDraw system, it is numbered (FastDraw 46) and color-coded to eliminate misuse. Spill-proof bottle helps prevent chemical contact with cleaning staff. Patented technology uses two metering tips for low-flow and high-flow dilution. Curved siphon tubes on the FastDraw Dispensers (not included) allow complete product evacuation, eliminating waste.