Learn More
Enterprise Technology Solutions Betco Corporation Carpet Cleaner, Encapsulation, 1 Gallon, Light Straw
SDP
Catalog No. 5000832505
Encompass_Preferred
FiberCAP MP contains advanced technology in encapsulation chemistry with polyhedral polymers to get all types of your commercial carpeting cleaner. The unique crystallization geometry of these polymers enhance soil entrapment and provides better release from the carpet fiber. Paired with the FiberCAP 20 Carpet Machine, FiberCAP MP leaves carpets cleaner longer. Low-moisture, multipurpose encapsulation cleaner dries fast after removing oily dirt and detergent residue. Routine maintenance is easy for quick removal of surface soils and spots without use of extraction machines. Dilution ratios depend on types of cleaning jobs: 1:10 for spot/traffic lane cleaning; 1:16 for interim/low-moisture cleaning; 1:64 for normal cleaning and 1:32 for heavy cleaning.
Catalog No. 50-008-32505 Supplier Enterprise Technology Solutions Supplier No. BET4200400
FiberCAP MP contains advanced technology in encapsulation chemistry with polyhedral polymers to get all types of your commercial carpeting cleaner. The unique crystallization geometry of these polymers enhance soil entrapment and provides better release from the carpet fiber. Paired with the FiberCAP 20 Carpet Machine, FiberCAP MP leaves carpets cleaner longer. Low-moisture, multipurpose encapsulation cleaner dries fast after removing oily dirt and detergent residue. Routine maintenance is easy for quick removal of surface soils and spots without use of extraction machines. Dilution ratios depend on types of cleaning jobs: 1:10 for spot/traffic lane cleaning; 1:16 for interim/low-moisture cleaning; 1:64 for normal cleaning and 1:32 for heavy cleaning.