Learn More
Enterprise Technology Solutions Betco Corporation Cleaner, f/Glass&Hard Surfaces, Conc, 2 L, Blue
SDP
Catalog No. 5000832221
Encompass_Preferred
Streak-free glass/surface cleaner cleans and polishes without scratching, streaking, fogging or clouding. It is concentrated for effective and economical use. It is the perfect product for glass, mirrors and many hard surfaces. Fragrance-free formula contains no butyl solvents or ammonia. Green Earth Glass Cleaner is certified for performance, concentration, minimized/recycled packaging and reduced toxicity by GreenSeal.org. As part of the FastDraw system, it is numbered (FastDraw 19) and color-coded to eliminate misuse. Spill-proof bottle helps prevent chemical contact with cleaning staff. Patented technology uses two metering tips for low-flow and high-flow dilution. Curved siphon tubes on the FastDraw Dispensers (not included) allow complete product evacuation, eliminating waste.
Catalog No. 50-008-32221 Supplier Enterprise Technology Solutions Supplier No. BET5354700
Streak-free glass/surface cleaner cleans and polishes without scratching, streaking, fogging or clouding. It is concentrated for effective and economical use. It is the perfect product for glass, mirrors and many hard surfaces. Fragrance-free formula contains no butyl solvents or ammonia. Green Earth Glass Cleaner is certified for performance, concentration, minimized/recycled packaging and reduced toxicity by GreenSeal.org. As part of the FastDraw system, it is numbered (FastDraw 19) and color-coded to eliminate misuse. Spill-proof bottle helps prevent chemical contact with cleaning staff. Patented technology uses two metering tips for low-flow and high-flow dilution. Curved siphon tubes on the FastDraw Dispensers (not included) allow complete product evacuation, eliminating waste.