Learn More
Enterprise Technology Solutions Betco Corporation Cleaner, f/Glass/Surfaces, Conc, 1/2 Gal (2L), 4/CT, Blue
SDP
Catalog No. 5000832348
Encompass_Preferred
Ammoniated glass/surface cleaner quickly penetrates dust, smoke, grease and grime on all glass and window surfaces. Deep Blue Concentrate is also excellent for cleaning stainless steel, Formica, plastic, porcelain and chrome. Formula contains ammonia for powerful cleaning on hard surfaces. Dilution ratio is 1:64. As part of the FastDraw system, it is numbered (FastDraw 23) and color-coded to eliminate misuse. Spill-proof bottle helps prevent chemical contact with cleaning staff. Patented technology uses two metering tips for low-flow and high-flow dilution. Curved siphon tubes on the FastDraw Dispensers (not included) allow complete product evacuation, eliminating waste.
Catalog No. 50-008-32348 Supplier Enterprise Technology Solutions Supplier No. BET1814700
Ammoniated glass/surface cleaner quickly penetrates dust, smoke, grease and grime on all glass and window surfaces. Deep Blue Concentrate is also excellent for cleaning stainless steel, Formica, plastic, porcelain and chrome. Formula contains ammonia for powerful cleaning on hard surfaces. Dilution ratio is 1:64. As part of the FastDraw system, it is numbered (FastDraw 23) and color-coded to eliminate misuse. Spill-proof bottle helps prevent chemical contact with cleaning staff. Patented technology uses two metering tips for low-flow and high-flow dilution. Curved siphon tubes on the FastDraw Dispensers (not included) allow complete product evacuation, eliminating waste.