Learn More
Enterprise Technology Solutions Betco Corporation Cleaner, f/Glass/Surfaces, FASTDRAW 5, 2L, Blue
SDP
Catalog No. 5000832220
Encompass_Preferred
This concentrated glass/surface cleaner quickly dissolves smudges, fingerprints, bug residues and other stubborn soils. It cleans and polishes glass without streaking, fogging or clouding. Dilute using a ratio of 2 oz. of Clear Image to 1 gallon of water (16 ml/1 liter). Formulated without ammonia, this nonflammable cleaner is pleasant to use and protects against resoiling. As part of the FastDraw system, the cleaner is numbered (FastDraw 5) and color-coded to eliminate cleaning staff misuse. NAFTA trilingual labels are suited to a multicultural workforce. Spill-proof bottle design eliminates the potential for chemical contact with cleaning staff. Patented technology uses two metering tips for low-flow and high-flow dilution. Metering insert reduces end-use cost per gallon by delivering accurate dilutions. Curved siphon tubes on the FastDraw Dispensers (not included) allow complete product evacuation, eliminating waste.
Catalog No. 50-008-32220 Supplier Enterprise Technology Solutions Supplier No. BET1994700
This concentrated glass/surface cleaner quickly dissolves smudges, fingerprints, bug residues and other stubborn soils. It cleans and polishes glass without streaking, fogging or clouding. Dilute using a ratio of 2 oz. of Clear Image to 1 gallon of water (16 ml/1 liter). Formulated without ammonia, this nonflammable cleaner is pleasant to use and protects against resoiling. As part of the FastDraw system, the cleaner is numbered (FastDraw 5) and color-coded to eliminate cleaning staff misuse. NAFTA trilingual labels are suited to a multicultural workforce. Spill-proof bottle design eliminates the potential for chemical contact with cleaning staff. Patented technology uses two metering tips for low-flow and high-flow dilution. Metering insert reduces end-use cost per gallon by delivering accurate dilutions. Curved siphon tubes on the FastDraw Dispensers (not included) allow complete product evacuation, eliminating waste.